Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011941.3 | NP_036071.3 | 25 | 3541 | ||
GenBank mRNA | AB029482.1 | - | 26 | 3355 | ||
AK122323.1 | - | 16 | 2240 | |||
BC011271.1 | - | 4 | 671 | |||
BC070449.1 | - | 25 | 3541 | 30543913 |