Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011998.4 | NP_036128.3 | 2 | 1125 | ||
GenBank mRNA | AF109155.1 | - | 2 | 1349 | ||
AI614882.1 | - | 1 | 101 | |||
AK009113.1 | - | 2 | 1126 | |||
BC057886.1 | - | 2 | 1145 | 4987327 |