Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013562.2 | NP_038590.2 | 11, 12 | 1500 | ||
GenBank mRNA | AK078888.1 | - | 1498 | |||
AK168682.1 | - | 11, 12 | 1496 | |||
AK213011.1 | - | 2, 3 | 146 | |||
AK215747.1 | - | 1, 2 | 70 | |||
BC037013.1 | - | 11, 12 | 1475 | |||
BC043723.1 | - | 11, 12 | 1467 | 5369100 | ||
V00756.1 | - | 1, 2 | 114 | |||
X17400.1 | - | 11, 12 | 1477 |