Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013724.2 | NP_038752.2 | 3, 4 | 506 | ||
GenBank mRNA | AB020741.1 | - | 3, 4 | 431 | ||
AB035267.1 | - | 3, 4 | 184 | |||
AK041377.1 | - | 3, 4 | 506 | |||
BC068311.1 | - | 3, 4 | 481 | |||
BC139029.1 | - | 3, 4 | 273 | 8862051 | ||
BC139031.1 | - | 3, 4 | 273 | 8862053 |