Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013791.2 | NP_038819.1 | 6 | 694 | ||
GenBank mRNA | AK037967.1 | - | 6 | 695 | ||
AK077541.1 | - | 6 | 690 | |||
AK136123.1 | - | 3 | 377 | |||
AK155457.1 | - | 6 | 675 | |||
AK164822.1 | - | 6 | 690 | |||
AK165239.1 | - | 6 | 694 | |||
BC013703.1 | - | 6 | 678 | 3668925 | ||
U72194.1 | - | 6 | 677 |