Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013748.3 | NP_038776.3 | 18 | 1207 | ||
GenBank mRNA | AB021220.1 | - | 18 | 1327 | ||
AF187819.1 | - | 18 | 1207 | |||
AK082826.1 | - | 14 | 945 | |||
BC125637.1 | - | 18 | 1204 | 40130152 | ||
BC125639.1 | - | 18 | 1204 | 40130154 |