Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015830.1 | NP_056645.1 | 1 | 648 | ||
GenBank mRNA | AK079422.1 | - | 2 | 881 | ||
AK143711.1 | - | 3 | 1287 | |||
AK170543.1 | - | 2 | 882 | |||
AK212570.1 | - | 1 | 89 | |||
BC058094.1 | - | 2 | 906 | 6824892 |