Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017476.2 | NP_059504.2 | 3, 4 | 207 | ||
GenBank mRNA | AB028921.1 | - | 3, 4 | 200 | ||
AK011325.1 | - | 3, 4 | 212 | |||
AK166526.1 | - | 3, 4 | 225 | |||
AK206808.1 | - | 2, 3 | 80 | |||
BC010401.1 | - | 3, 4 | 257 | |||
BC046801.1 | - | 3, 4 | 196 | 6403966 | ||
BC051015.1 | - | 3, 4 | 207 |