Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017366.3 | NP_059062.1 | 4 | 353 | ||
GenBank mRNA | AK076037.1 | - | 4 | 314 | ||
AU067294.1 | - | 4 | 353 | |||
BC026559.1 | - | 4 | 332 | 5037878 | ||
Z71189.1 | - | 4 | 274 |