Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018817.2 | NP_061287.2 | 2 | 570 | ||
GenBank mRNA | AF088884.1 | - | 2 | 550 | ||
AK035854.1 | - | 2 | 547 | |||
AK042332.1 | - | 2 | 549 | |||
AK077878.1 | - | 2 | 548 | |||
AK083488.1 | - | 2 | 548 | |||
AK146767.1 | - | 2 | 564 | |||
AK154020.1 | - | 2 | 553 | |||
AK161570.1 | - | 2 | 490 | |||
AK169465.1 | - | 2 | 561 | |||
BC029078.1 | - | 2 | 505 | 3992325 |