Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018878.3 | NP_061366.2 | 1, 2 | 360 | ||
GenBank mRNA | AF104261.1 | - | 1, 2 | 360 | ||
AK144606.1 | - | 1, 2 | 222 | |||
AK148141.1 | - | 1, 2 | 209 | |||
AK198400.1 | - | 1, 2 | 31 | |||
BC066014.1 | - | 1, 2 | 149 | 5704566 |