Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019505.3 | NP_062378.1 | 1, 2 | 826 | ||
GenBank mRNA | AF136744.1 | - | 1, 2 | 463 | ||
AK044596.1 | - | 1562 | ||||
AK049335.1 | - | 1, 2 | 626 | |||
AK147841.1 | - | 1, 2 | 827 | |||
BB635763.1 | - | 1, 2 | 230 | |||
BC055867.1 | - | 1, 2 | 759 |