Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301671.1 | NP_001288600.1 | 4 | 592 | ||
NM_001301673.1 | NP_001288602.1 | 4 | 475 | |||
NM_019929.4 | NP_064313.1 | 4 | 466 | |||
GenBank mRNA | AA105565.1 | - | 3 | 196 | ||
AF063847.1 | - | 4 | 293 | |||
AK012742.1 | - | 4 | 341 | |||
AK013019.1 | - | 4 | 335 | |||
AK148306.1 | - | 4 | 330 | |||
AK150063.1 | - | 4 | 339 | |||
AK150200.1 | - | 4 | 339 | |||
AK160169.1 | - | 4 | 338 | |||
AK162035.1 | - | 4 | 339 | |||
AK167207.1 | - | 4 | 307 | |||
AK168988.1 | - | 4 | 330 | |||
AK170896.1 | - | 4 | 524 | |||
AK171169.1 | - | 4 | 524 | |||
BC115488.1 | - | 4 | 273 | 40100722 | ||
BC115489.1 | - | 4 | 273 | 40100723 | ||
CJ069985.1 | - | 4 | 377 | |||
CJ159246.1 | - | 4 | 383 | |||
CK619326.1 | - | 5 | 434 |