Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020486.2 | NP_065232.1 | 5, 6 | 652 | ||
GenBank mRNA | AB035511.1 | - | 5, 6 | 613 | ||
AF109160.1 | - | 5, 6 | 652 | |||
AF221507.1 | - | 5, 6 | 602 | |||
AF346663.1 | - | 5, 6 | 590 | |||
AK040564.1 | - | 5, 6 | 636 | |||
AK054001.1 | - | 5, 6 | 633 | |||
AK075704.1 | - | 5, 6 | 617 | |||
AK158439.1 | - | 5, 6 | 614 | |||
AK219904.1 | - | 1, 2 | 122 | |||
BC004826.1 | - | 5, 6 | 636 | 2616148 |