Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021305.3 | NP_067280.1 | 4, 5 | 318 | ||
GenBank mRNA | AF145254.1 | - | 4, 5 | 254 | ||
AF222748.1 | - | 4, 5 | 255 | |||
AK017615.1 | - | 4, 5 | 300 | |||
AK077701.1 | - | 4, 5 | 318 | |||
AK136447.1 | - | 4, 5 | 297 | |||
AK160602.1 | - | 4, 5 | 304 | |||
AK161316.1 | - | 4, 5 | 397 | |||
BC005458.1 | - | 4, 5 | 261 | 3494001 |