Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199060.1 | NP_001185989.1 | 5 | 783 | ||
NM_001199061.1 | NP_001185990.1 | 5 | 515 | |||
NM_021312.5 | NP_067287.1 | 4 | 605 | |||
GenBank mRNA | AB041608.1 | - | 5 | 784 | ||
AF239765.1 | - | 4 | 261 | |||
AK007500.1 | - | 4 | 382 | |||
AK012022.1 | - | 4 | 359 | |||
AK016611.1 | - | 5 | 459 | |||
AK031436.1 | - | 4 | 368 | |||
AK149085.1 | - | 4 | 287 | |||
AK161299.1 | - | 5 | 528 | |||
AK213584.1 | - | 3 | 154 | |||
AY059431.1 | - | 4 | 296 | |||
BC004748.1 | - | 4 | 439 | 3583955 | ||
BC052386.1 | - | 4 | 576 | 6418498 | ||
BC096651.1 | - | 4 | 350 | 1514703 | ||
CX564842.1 | - | 4 | 606 |