Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANAPC5 SIRNA Assays ›
Gene Symbol:
Anapc5
Gene Name:
anaphase-promoting complex subunit 5
Gene Aliases:
2510006G12Rik, AA408751, AA536819, AA986414, APC5, Anpc5
Chromosome Location:
Chr.5: 122787459 - 122821342 on Build GRCm38
UniGene ID:
Mm.332667
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
75864
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Anapc5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042491.2
NP_001035956.1
3
471
NM_001289517.1
NP_001276446.1
3
471
NM_001289518.1
NP_001276447.1
3
471
NM_001289519.1
NP_001276448.1
3
471
NM_001289520.1
NP_001276449.1
3
471
NM_021505.3
NP_067480.2
3
471
GenBank mRNA
AB041593.1
-
3
414
AK013978.1
-
3
395
AK031432.1
-
3
396
AK049719.1
-
3
425
AK075969.1
-
3
410
AK088327.1
-
3
434
AK147951.1
-
3
410
AK148679.1
-
3
426
AK151314.1
-
3
417
AK151905.1
-
3
417
AK152528.1
-
3
387
AK152981.1
-
3
417
AK153373.1
-
3
417
AK153422.1
-
3
417
AK159204.1
-
3
417
AK159713.1
-
3
410
AK159916.1
-
3
410
AK167561.1
-
3
410
AK168135.1
-
3
395
AK168737.1
-
3
411
AK168814.1
-
3
410
AK169196.1
-
3
434
AK180837.1
-
3
144
AK182300.1
-
3
144
AK192754.1
-
3
167
AK194108.1
-
3
136
AK197886.1
-
3
143
AK203216.1
-
2
120
AK215079.1
-
3
144
AK217317.1
-
3
149
BC010339.1
-
3
387
2648173
BC046566.1
-
3
450
2650790
BC046804.1
-
3
408
6404923
BQ950394.1
-
3
471
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
mitotic nuclear division
cell division
protein K11-linked ubiquitination
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0