Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021539.4 | NP_067514.2 | 2, 3 | 338 | ||
GenBank mRNA | AF033188.1 | - | 1, 2 | 182 | ||
AF072881.1 | - | 2, 3 | 259 | |||
AK136416.1 | - | 1, 2 | 238 | |||
AK157999.1 | - | 2, 3 | 297 | |||
BC055100.1 | - | 2, 3 | 338 | 6589250 |