Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022022.3 | NP_071305.2 | 6 | 1331 | ||
GenBank mRNA | AF260926.1 | - | 6 | 1446 | ||
AK016689.1 | - | 6 | 1332 | |||
AK029914.1 | - | 6 | 936 | |||
AK083409.1 | - | 6 | 933 | |||
AK122345.1 | - | 6 | 842 | |||
BC067402.1 | - | 6 | 1078 | 30099547 | ||
BC075620.1 | - | 6 | 1292 | 6405514 |