Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258459.1 | NP_001245388.1 | 6, 7 | 674 | ||
NM_001258460.1 | NP_001245389.1 | 5, 6 | 551 | |||
NM_006213.4 | NP_006204.1 | 5, 6 | 578 | |||
NR_047689.1 | 4, 5 | 523 | ||||
XM_005271772.4 | XP_005271829.1 | 3, 4 | 795 | |||
XM_017012324.1 | XP_016867813.1 | 5, 6 | 578 | |||
XM_017012325.1 | XP_016867814.1 | 2, 3 | 239 | |||
XM_017012326.1 | XP_016867815.1 | 3, 4 | 798 | |||
XM_017012327.1 | XP_016867816.1 | 2, 3 | 207 | |||
GenBank mRNA | AK293180.1 | - | 6, 7 | 629 | ||
AK299429.1 | - | 4, 5 | 479 | |||
AK300928.1 | - | 5, 6 | 503 | |||
AK312851.1 | - | 5, 6 | 578 | |||
BC051327.1 | - | 5, 6 | 516 | 6277230 | ||
BC069655.1 | - | 5, 6 | 503 | 7262499 | ||
BC069679.1 | - | 5, 6 | 494 | 7262511 | ||
BC069738.1 | - | 5, 6 | 495 | 7262523 | ||
BC069754.1 | - | 5, 6 | 495 | 7262535 | ||
BC074753.2 | - | 4, 5 | 422 | 30915212 | ||
X80590.1 | - | 5, 6 | 502 |