Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024221.3 | NP_077183.1 | 3 | 192 | ||
GenBank mRNA | AK011810.1 | - | 156 | |||
AK014180.1 | - | 3 | 148 | |||
AK014918.1 | - | 3 | 193 | |||
AK084507.1 | - | 2 | 1383 | |||
AK153058.1 | - | 3 | 146 | |||
AK163596.1 | - | 1 | 1469 | |||
AK166631.1 | - | 3 | 194 | |||
AK167798.1 | - | 3 | 149 | |||
BC019512.1 | - | 3 | 165 | 4210204 | ||
BC094468.1 | - | 3 | 138 | 5011266 | ||
CJ159324.1 | - | 3 | 196 |