Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024211.3 | NP_077173.1 | 2, 3 | 419 | ||
GenBank mRNA | AI931249.1 | - | 2, 3 | 380 | ||
AK003957.1 | - | 1, 2 | 58 | |||
AK009487.1 | - | 2, 3 | 365 | |||
AK009824.1 | - | 2, 3 | 319 | |||
AK168951.1 | - | 2, 3 | 344 | |||
AK181540.1 | - | 1, 2 | 101 | |||
AK187524.1 | - | 1, 2 | 101 | |||
AK202646.1 | - | 1, 2 | 101 | |||
AK212146.1 | - | 1, 2 | 91 | |||
AK213879.1 | - | 1, 2 | 101 | |||
AK215545.1 | - | 1, 2 | 91 | |||
BC003455.1 | - | 2, 3 | 395 | 2648336 | ||
BC019631.1 | - | 2, 3 | 373 | 4023179 |