Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039147.2 | NP_001034236.1 | 3 | 361 | ||
NM_024431.3 | NP_077751.1 | 3 | 361 | |||
GenBank mRNA | AB042241.1 | - | 3 | 210 | ||
AF319620.1 | - | 3 | 209 | |||
AF319621.1 | - | 3 | 209 | |||
AK011249.1 | - | 3 | 238 | |||
AK011441.1 | - | 3 | 133 | |||
AK016414.1 | - | 3 | 328 | |||
AK146210.1 | - | 3 | 196 | |||
AK146785.1 | - | 2 | 91 | |||
AK150925.1 | - | 3 | 162 | |||
AK152119.1 | - | 3 | 138 | |||
AK167079.1 | - | 3 | 120 | |||
AK168327.1 | - | 3 | 196 | |||
AK168585.1 | - | 3 | 141 | |||
AK168608.1 | - | 3 | 143 | |||
AK168674.1 | - | 3 | 192 | |||
AK169065.1 | - | 3 | 193 | |||
AK182816.1 | - | 3 | 133 | |||
AK187780.1 | - | 3 | 133 | |||
AK190529.1 | - | 3 | 133 | |||
AK198286.1 | - | 3 | 133 | |||
AK207086.1 | - | 3 | 133 | |||
AK207233.1 | - | 3 | 133 | |||
AK211227.1 | - | 3 | 133 | |||
AK211776.1 | - | 3 | 132 | |||
AK212639.1 | - | 3 | 133 | |||
AK212964.1 | - | 3 | 133 | |||
AK213758.1 | - | 3 | 133 | |||
AK214297.1 | - | 3 | 133 | |||
AK214633.1 | - | 3 | 133 | |||
AK215083.1 | - | 3 | 133 | |||
AK215514.1 | - | 3 | 129 | |||
AK218105.1 | - | 3 | 133 | |||
AK218718.1 | - | 3 | 133 | |||
AK218976.1 | - | 3 | 133 | |||
AK220441.1 | - | 3 | 181 | |||
BC003894.1 | - | 3 | 213 | 3593982 | ||
BC017619.1 | - | 3 | 213 | 4923923 | ||
BC083094.1 | - | 3 | 146 | |||
BC083118.1 | - | 3 | 222 | 5371036 | ||
BC085103.1 | - | 3 | 211 | 6490291 | ||
BC092293.1 | - | 3 | 225 | 5068516 | ||
BC103783.1 | - | 3 | 225 | 6529024 |