Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164708.1 | NP_001158180.1 | 2 | 332 | ||
NM_024441.3 | NP_077761.3 | 2 | 614 | |||
GenBank mRNA | AK012780.1 | - | 2 | 333 | ||
BC064051.1 | - | 2 | 553 | 948723 | ||
BU704695.1 | - | 86 |