Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242413.2 | NP_001229342.1 | 2 | 171 | ||
NM_001282644.1 | NP_001269573.1 | 2 | 171 | |||
NM_001282645.1 | NP_001269574.1 | 2 | 171 | |||
NM_001323265.1 | NP_001310194.1 | 2 | 226 | |||
NM_001323266.1 | NP_001310195.1 | 2 | 171 | |||
NM_001323267.1 | NP_001310196.1 | 2 | 171 | |||
NM_006257.4 | NP_006248.1 | 2 | 171 | |||
XM_005252496.4 | XP_005252553.1 | 2 | 271 | |||
XM_005252497.4 | XP_005252554.1 | 2 | 273 | |||
XM_006717465.3 | XP_006717528.1 | 4 | 2666 | |||
XM_017016410.1 | XP_016871899.1 | 3 | 2577 | |||
XM_017016411.1 | XP_016871900.1 | 4 | 2666 | |||
GenBank mRNA | AK293935.1 | - | 2 | 109 | ||
AK308099.1 | - | 2 | 146 | |||
AK312411.1 | - | 2 | 121 | |||
AU099414.1 | - | 2 | 146 | |||
AY702977.1 | - | 1 | 71 | |||
BC101465.1 | - | 1 | 90 | 8068971 | ||
BC113359.1 | - | 1 | 90 | 8322411 | ||
BX647657.1 | - | 1 | 255 | |||
L01087.1 | - | 2 | 165 | |||
L07032.1 | - | 1 | 92 |