Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025641.3 | NP_079917.1 | 3 | 298 | ||
GenBank mRNA | AK003266.1 | - | 3 | 263 | ||
AK003539.1 | - | 3 | 241 | |||
AK011734.1 | - | 660 | ||||
AK012552.1 | - | 3 | 276 | |||
AK018712.1 | - | 3 | 276 | |||
AK019085.1 | - | 3 | 155 | |||
AK028137.1 | - | 1 | 40 | |||
AK054005.1 | - | 3 | 264 | |||
AK088185.1 | - | 3 | 263 | |||
BC011388.1 | - | 3 | 242 | 3597862 | ||
BY138998.1 | - | 3 | 298 | |||
BY702808.1 | - | 3 | 271 |