Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025611.5 | NP_079887.3 | 24 | 4912 | ||
GenBank mRNA | AK008655.1 | - | 1 | 100 | ||
AK010907.1 | - | 3 | 437 | |||
AK147156.1 | - | 24 | 4670 | |||
AK147905.1 | - | 24 | 4632 | |||
AK170808.1 | - | 24 | 4685 | |||
AK172882.1 | - | 23 | 3985 | |||
BC019645.1 | - | 14 | 2200 | |||
BC024345.1 | - | 3 | 704 | |||
BC026946.1 | - | 6 | 1020 | |||
BC059865.1 | - | 20 | 3425 | |||
BC065096.1 | - | 23 | 4399 | |||
BC100544.1 | - | 3 | 548 | |||
BC138998.1 | - | 24 | 4850 | 8862020 | ||
BC139000.1 | - | 24 | 4850 | 8862022 |