Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025624.2 | NP_079900.1 | 5, 6 | 414 | ||
GenBank mRNA | AK003439.1 | - | 5, 6 | 407 | ||
AK011115.1 | - | 5, 6 | 415 | |||
AK012455.1 | - | 5, 6 | 376 | |||
AK013934.1 | - | 5, 6 | 408 | |||
AK014296.1 | - | 5, 6 | 378 | |||
AK132041.1 | - | 391 | ||||
AK167958.1 | - | 5, 6 | 415 | |||
BC027016.1 | - | 5, 6 | 405 | 5375452 |