Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025836.3 | NP_080112.1 | 5 | 449 | ||
GenBank mRNA | AK004970.1 | - | 5 | 450 | ||
AK012517.1 | - | 5 | 450 | |||
AK049537.1 | - | 5 | 450 | |||
AK050140.1 | - | 5 | 451 | |||
AK132313.1 | - | 450 | ||||
AK167214.1 | - | 5 | 448 | |||
AK197135.1 | - | 3 | 207 | |||
BC011116.1 | - | 5 | 445 | 4023354 | ||
DQ257436.1 | - | 5 | 408 |