Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000028.2 | NP_000019.2 | 6 | 1276 | ||
NM_000642.2 | NP_000633.2 | 6 | 1198 | |||
NM_000643.2 | NP_000634.2 | 6 | 996 | |||
NM_000644.2 | NP_000635.2 | 6 | 936 | |||
NM_000645.2 | NP_000636.2 | 4 | 1009 | |||
NM_000646.2 | NP_000637.2 | 6 | 1009 | |||
XM_005270557.2 | XP_005270614.1 | 6 | 891 | |||
GenBank mRNA | AB208867.1 | - | 5 | 802 | ||
AB621804.1 | - | 6 | 1146 | |||
BC078663.1 | - | 6 | 918 | 30350685 | ||
M85168.1 | - | 4 | 1011 | |||
U84007.1 | - | 6 | 1198 | |||
U84008.1 | - | 6 | 936 | |||
U84009.1 | - | 6 | 996 | |||
U84010.1 | - | 6 | 1276 | |||
U84011.1 | - | 6 | 1009 |