Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026036.3 | NP_080312.1 | 4 | 678 | ||
GenBank mRNA | AK012941.1 | - | 4 | 679 | ||
AK030885.1 | - | 4 | 658 | |||
AK128960.1 | - | 4 | 664 | |||
AK145743.1 | - | 4 | 659 | |||
AK150775.1 | - | 4 | 677 | |||
AK151092.1 | - | 4 | 677 | |||
AK151249.1 | - | 1 | 197 | |||
AK151431.1 | - | 4 | 677 | |||
AK152448.1 | - | 4 | 680 | |||
AK152723.1 | - | 4 | 689 | |||
AK152817.1 | - | 4 | 689 | |||
AK153092.1 | - | 4 | 677 | |||
AK154481.1 | - | 4 | 665 | |||
AK159439.1 | - | 4 | 648 | |||
AK159735.1 | - | 4 | 673 | |||
AK165948.1 | - | 4 | 647 | |||
AK167499.1 | - | 4 | 665 | |||
AK167854.1 | - | 4 | 670 | |||
AK170905.1 | - | 4 | 680 | |||
AK171394.1 | - | 4 | 709 | |||
AY241868.1 | - | 4 | 715 | |||
BC007194.1 | - | 2 | 326 | |||
BC027248.1 | - | 4 | 650 | 2811938 |