Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026278.3 | NP_080554.1 | 2, 3 | 318 | ||
GenBank mRNA | AK006731.1 | - | 515 | |||
AK007202.1 | - | 2, 3 | 310 | |||
AK015594.1 | - | 2, 3 | 311 | |||
AK016626.1 | - | 323 | ||||
BC051150.1 | - | 2, 3 | 318 | 6772814 |