Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177576.1 | NP_001171047.1 | 4 | 406 | ||
NM_026646.3 | NP_080922.1 | 4 | 648 | |||
GenBank mRNA | AK010193.1 | - | 4 | 394 | ||
AK018760.1 | - | 4 | 412 | |||
AK046874.1 | - | 4 | 393 | |||
AK048389.1 | - | 4 | 377 | |||
AK049313.1 | - | 4 | 377 | |||
AK155276.1 | - | 4 | 400 | |||
AK158985.1 | - | 1 | 1748 | |||
AK162400.1 | - | 4 | 359 | |||
BC037949.1 | - | 4 | 405 | 5367108 | ||
BC050887.1 | - | 4 | 601 |