Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dopey2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026700.2 | NP_080976.1 | 3 | 498 | ||
NM_027293.1 | NP_081569.1 | 3 | 498 | |||
GenBank mRNA | AK002808.1 | - | 3 | 428 | ||
AK147255.1 | - | 3 | 476 | |||
AK147536.1 | - | 2 | 518 | |||
AK164839.1 | - | 3 | 475 |