Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027019.3 | NP_081295.2 | 6 | 626 | ||
GenBank mRNA | AB067773.1 | - | 6 | 715 | ||
AK005883.1 | - | 6 | 737 | |||
BC104105.1 | - | 6 | 626 | 40043157 | ||
BC104106.1 | - | 6 | 626 | 40043160 | ||
BC106806.1 | - | 6 | 626 | 40042632 |