Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026906.3 | NP_081182.2 | 7 | 1050 | ||
NM_054092.2 | NP_473433.2 | 7 | 975 | |||
GenBank mRNA | AF456464.1 | - | 6 | 835 | ||
AK005389.1 | - | 7 | 973 | |||
AY034574.1 | - | 7 | 989 | |||
BC099388.1 | - | 7 | 944 | 30786828 |