Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027036.3 | NP_081312.2 | 1 | 505 | ||
GenBank mRNA | AK005588.1 | - | 1 | 504 | ||
BC048744.1 | - | 2 | 465 | |||
BC061030.1 | - | 1 | 501 | 6775221 | ||
BC087910.1 | - | 2 | 391 | |||
BC087934.1 | - | 2 | 461 | |||
CA465179.1 | - | 1 | 504 | |||
CN840046.1 | - | 1 | 508 |