Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027122.3 | NP_081398.2 | 1 | 267 | ||
GenBank mRNA | AK009269.1 | - | 1 | 346 | ||
AK089539.1 | - | 1 | 268 | |||
BC019171.1 | - | 1 | 294 | 5003354 | ||
BC085305.1 | - | 1 | 331 | 6399056 |