Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eqtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290623.1 | NP_001277552.1 | 7 | 633 | ||
NM_027089.4 | NP_081365.2 | 7 | 633 | |||
GenBank mRNA | AB438105.1 | - | 7 | 542 | ||
AB438106.1 | - | 7 | 542 | |||
AK006446.1 | - | 7 | 584 | |||
AK016738.1 | - | 7 | 604 | |||
BC089517.1 | - | 7 | 600 | 6740450 | ||
BE632290.1 | - | 3 | 152 | |||
DQ336137.1 | - | 7 | 584 | |||
EU828781.1 | - | 7 | 618 | |||
EU828782.1 | - | 7 | 618 |