Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027582.3 | NP_081858.2 | 3 | 412 | ||
GenBank mRNA | AK014937.1 | - | 2 | 291 | ||
AK135492.1 | - | 3 | 415 | |||
BC051128.1 | - | 3 | 412 | 6707386 |