Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Telo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163661.1 | NP_001157133.1 | 2 | 646 | ||
NM_027880.2 | NP_082156.2 | 2 | 557 | |||
GenBank mRNA | AK004582.1 | - | 2 | 427 | ||
AK028658.1 | - | 2 | 411 | |||
AK050773.1 | - | 2 | 377 | |||
AK084830.1 | - | 2 | 427 | |||
AK129189.1 | - | 2 | 595 | |||
AK141388.1 | - | 2 | 623 | |||
AK143827.1 | - | 2 | 647 | |||
AK146963.1 | - | 2 | 433 | |||
AK154727.1 | - | 2 | 459 | |||
BC011077.1 | - | 2 | 407 | 3984911 | ||
BC013127.1 | - | 2 | 633 | |||
BC080713.1 | - | 2 | 586 |