Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF356878.1 | - | 4 | 518 | ||
AF356879.1 | - | 3 | 377 | |||
AF375479.1 | - | 2 | 329 | |||
AK002247.1 | - | 4 | 517 | |||
AK143782.1 | - | 4 | 520 | |||
AY169234.1 | - | 4 | 517 | |||
BC010795.1 | - | 2 | 738 | 4221711 |