Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027990.3 | NP_082266.1 | 10 | 1117 | ||
GenBank mRNA | AK009282.1 | - | 10 | 1117 | ||
BC126943.1 | - | 7 | 861 | 40090023 |