Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 5 | 636 | ||
GenBank mRNA | AK005460.1 | - | 5 | 638 | ||
AK051121.1 | - | 4 | 854 | |||
AK051628.1 | - | 5 | 654 | |||
AK081672.1 | - | 5 | 552 | |||
AK159555.1 | - | 5 | 681 | |||
AK159871.1 | - | 5 | 666 | |||
AK166578.1 | - | 5 | 644 | |||
AK167201.1 | - | 5 | 657 | |||
AK167613.1 | - | 5 | 654 | |||
AK169021.1 | - | 5 | 661 | |||
AK169098.1 | - | 5 | 681 | |||
AK169410.1 | - | 5 | 611 | |||
AK179383.1 | - | 5 | 368 | |||
AK180041.1 | - | 4 | 386 | |||
AK184231.1 | - | 5 | 372 | |||
AK190584.1 | - | 4 | 381 | |||
AK208457.1 | - | 4 | 336 | |||
AK212516.1 | - | 4 | 285 | |||
AK215736.1 | - | 4 | 346 | |||
AK216318.1 | - | 5 | 367 | |||
BC039643.1 | - | 1 | 31 | |||
BC055711.1 | - | 5 | 637 | 6415164 | ||
BC085268.1 | - | 5 | 414 | 6487559 |