Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028123.3 | NP_082399.3 | 15 | 2588 | ||
GenBank mRNA | AK012071.1 | - | 7 | 1811 | ||
AK076213.1 | - | 15 | 2531 | |||
AK077836.1 | - | 15 | 2531 | |||
AK145242.1 | - | 15 | 2569 | |||
AK167716.1 | - | 15 | 2544 | |||
BC005744.1 | - | 15 | 2407 | |||
BC099673.1 | - | 16 | 2525 | 4936456 | ||
BC100391.1 | - | 16 | 2622 |