Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028301.4 | NP_082577.2 | 11, 12 | 1520 | ||
GenBank mRNA | AK012494.1 | - | 11, 12 | 1469 | ||
AK129471.1 | - | 10, 11 | 2549 | |||
BC022691.1 | - | 3, 4 | 376 | |||
BC039644.1 | - | 4, 5 | 473 | |||
BC050929.1 | - | 11, 12 | 1456 | 6807194 |