Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028627.2 | NP_082903.2 | 18 | 3790 | ||
GenBank mRNA | AK003542.1 | - | 2 | 766 | ||
AK158685.1 | - | 8 | 1598 | |||
AK158851.1 | - | 8 | 1722 | |||
AK220438.1 | - | 13 | 3908 | |||
BC043135.1 | - | 14 | 2620 | |||
BC057353.1 | - | 18 | 3792 | |||
BC058352.1 | - | 4 | 1165 | |||
BC069847.1 | - | 1 | 487 |