Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028716.4 | NP_082992.1 | 12 | 1207 | ||
GenBank mRNA | AK014380.1 | - | 12 | 1208 | ||
BC027776.1 | - | 12 | 1202 | 5352251 |