Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028880.3 | NP_083156.2 | 2 | 921 | ||
GenBank mRNA | AK014545.1 | - | 2 | 924 | ||
AK053345.1 | - | 2 | 911 | |||
AK053364.1 | - | 2 | 545 | |||
AK080836.1 | - | 2 | 952 | |||
AK136857.1 | - | 1 | 420 | |||
AY182025.1 | - | 1 | 356 | |||
BC027803.1 | - | 2 | 803 | 5321979 |