Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028801.2 | NP_083077.2 | 25, 26 | 3368 | ||
GenBank mRNA | AJ311906.1 | - | 3370 | |||
AK037730.1 | - | 25, 26 | 3369 |